Merck
  • Methionine oxidation activates a transcription factor in response to oxidative stress.
Methionine oxidation activates a transcription factor in response to oxidative stress.
Proceedings of the National Academy of Sciences of the United States of America (2013-05-22)
Adrian Drazic, Haruko Miura, Jirka Peschek, Yan Le, Nina C Bach, Thomas Kriehuber, Jeannette Winter
ABSTRACT
Oxidant-mediated antibacterial response systems are broadly used to control bacterial proliferation. Hypochlorite (HOCl) is an important component of the innate immune system produced in neutrophils and specific epithelia. Its antimicrobial activity is due to damaging cellular macromolecules. Little is known about how bacteria escape HOCl-inflicted damage. Recently, the transcription factor YjiE was identified that specifically protects Escherichia coli from HOCl killing. According to its function, YjiE is now renamed HypT (hypochlorite-responsive transcription factor). Here we unravel that HypT is activated by methionine oxidation to methionine sulfoxide. Interestingly, so far only inactivation of cellular proteins by methionine oxidation has been reported. Mutational analysis revealed three methionines that are essential to confer HOCl resistance. Their simultaneous substitution by glutamine, mimicking the methionine sulfoxide state, increased the viability of E. coli cells upon HOCl stress. Triple glutamine substitution generates a constitutively active HypT that regulates target genes independently of HOCl stress and permanently down-regulates intracellular iron levels. Inactivation of HypT depends on the methionine sulfoxide reductases A/B. Thus, microbial protection mechanisms have evolved along the evolution of antimicrobial control systems, allowing bacteria to survive within the host environment.
Sigma-Aldrich
DL-Methionine, 99%, FCC, FG
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
DL-Methionine, ≥99%
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
DL-Methionine, ≥99.0% (NT)
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)
SAFC
L-Methionine
Sigma-Aldrich
DL-Methionine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Supelco
L-Methionine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Methionine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Methionine, Pharmaceutical Secondary Standard; Certified Reference Material