Merck
  • Cortical inhibition reduces information redundancy at presentation of communication sounds in the primary auditory cortex.
Cortical inhibition reduces information redundancy at presentation of communication sounds in the primary auditory cortex.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2013-06-28)
Quentin Gaucher, Chloé Huetz, Boris Gourévitch, Jean-Marc Edeline
ABSTRACT
In all sensory modalities, intracortical inhibition shapes the functional properties of cortical neurons but also influences the responses to natural stimuli. Studies performed in various species have revealed that auditory cortex neurons respond to conspecific vocalizations by temporal spike patterns displaying a high trial-to-trial reliability, which might result from precise timing between excitation and inhibition. Studying the guinea pig auditory cortex, we show that partial blockage of GABAA receptors by gabazine (GBZ) application (10 μm, a concentration that promotes expansion of cortical receptive fields) increased the evoked firing rate and the spike-timing reliability during presentation of communication sounds (conspecific and heterospecific vocalizations), whereas GABAB receptor antagonists [10 μm saclofen; 10-50 μm CGP55845 (p-3-aminopropyl-p-diethoxymethyl phosphoric acid)] had nonsignificant effects. Computing mutual information (MI) from the responses to vocalizations using either the evoked firing rate or the temporal spike patterns revealed that GBZ application increased the MI derived from the activity of single cortical site but did not change the MI derived from population activity. In addition, quantification of information redundancy showed that GBZ significantly increased redundancy at the population level. This result suggests that a potential role of intracortical inhibition is to reduce information redundancy during the processing of natural stimuli.
Sigma-Aldrich
CGP 55845 hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Saclofen, solid
Sigma-Aldrich
(±)-Baclofen, ≥98% (HPLC), solid
Sigma-Aldrich
SR-95531, ≥98% (HPLC), powder