Merck
  • Effects of topical antiglaucoma eye drops on prostaglandin E(2)-induced aqueous flare elevation in pigmented rabbits.
Effects of topical antiglaucoma eye drops on prostaglandin E(2)-induced aqueous flare elevation in pigmented rabbits.
Investigative ophthalmology & visual science (2002-03-30)
Yoriko Hayasaka, Seiji Hayasaka, Xue-Yun Zhang, Yasunori Nagaki
ABSTRACT
To evaluate the role of topical instillation of some antiglaucoma agents on experimental elevation of aqueous flare induced by prostaglandin E(2) (PGE(2)) in pigmented rabbits. Transcorneal diffusion of PGE(2) (25 microg/mL or 7.09 x 10(-2) mM) with the use of a glass cylinder was achieved to produce aqueous flare elevation in pigmented rabbits. An antiglaucoma agent was topically administered before application of PGE(2). Aqueous flare was measured with a laser flare cell meter. A single instillation of apraclonidine 1.15%, two instillations of epinephrine 1.25%, two instillations of dipivefrin 0.1%, and two instillations and one instillation of dipivefrin 0.04% eye drops inhibited 98%, 96%, 87%, 73%, and 47% of PGE(2)-induced aqueous flare elevation, respectively. Timolol 0.5%, nipradilol 0.25%, dorzolamide 1%, and pilocarpine 2% eye drops had no effects on the increase of PGE(2)-induced flare. Apraclonidine, epinephrine, and dipivefrin eye drops inhibit PGE(2)-induced elevation of aqueous flare in pigmented rabbits.
Product Description
Dipivefrine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Dipivefrine for system suitability, European Pharmacopoeia (EP) Reference Standard