Merck
  • Viral induction and targeted inhibition of galectin-1 in EBV+ posttransplant lymphoproliferative disorders.
Viral induction and targeted inhibition of galectin-1 in EBV+ posttransplant lymphoproliferative disorders.
Blood (2011-02-09)
Jing Ouyang, Przemyslaw Juszczynski, Scott J Rodig, Michael R Green, Evan O'Donnell, Treeve Currie, Myriam Armant, Kunihiko Takeyama, Stefano Monti, Gabriel A Rabinovich, Jerome Ritz, Jeffery L Kutok, Margaret A Shipp
ABSTRACT
Posttransplant lymphoproliferative disorders (PTLDs) are potentially fatal, EBV-driven B-cell malignancies that develop in immunocompromised solid organ or hematopoietic stem cell recipients. In PTLD, the expression of EBV proteins, including latent membrane protein 1 (LMP1) and LMP2A, viral immune evasion strategies, and impaired host immune surveillance foster the proliferation of EBV-transformed B cells. Current PTLD treatment strategies include reduction of immunosuppression, which increases the risk of graft rejection, anti-CD20 treatment, combination chemotherapy, and administration of EBV-specific cytotoxic T cells. In the present study, we report that EBV-transformed lymphoblastoid B-cell lines (LCLs) and primary PTLDs overexpress galectin-1 (Gal1), a carbohydrate-binding lectin that induces tolerogenic dendritic cells and triggers the selective apoptosis of CD4(+) Th1 and Th17 cells and cytotoxic T cells. In transcriptional reporter assays, LMP2A and LMP1 each increased Gal1-driven luciferase expression, and the combination of LMP2A and LMP1 was additive. In addition, small interfering RNA (siRNA)-mediated depletion of LMP2A decreased Gal1 protein abundance in EBV-transformed LCLs. Gal1 expression in LCLs was dependent on both activating protein 1 (AP-1) and PI3K. A newly developed neutralizing Gal1 mAb selectively inhibited Gal1-mediated apoptosis of EBV-specific CD8(+) T cells. Given the tolerogenic and immunosuppressive function of Gal1, antibody-mediated Gal1 neutralization may represent a novel immunotherapeutic strategy for PTLD and other Gal1-expressing tumors.
Sigma-Aldrich
LY 294002, LY294002, CAS 154447-36-6, is a cell-permeable, potent, reversible, and specific inhibitor of PI 3-kinase ((IC50 = 1.4 µM). Acts on the ATP-binding site.
Millipore
JAK Inhibitor I, JAK Inhibitor I, CAS 457081-03-7, is a potent, reversible, cell-permeable, and ATP-competitive inhibitor of JAK 1 (IC50 = 15 nM), JAK2 (IC50 = 1 nM), JAK3 (Ki = 5 nM) and Tyk2 (IC50 = 1 nM).