Merck
A new immunochemical assay for biotin.
Clinica chimica acta; international journal of clinical chemistry (1991-10-31)
L P Thuy, L Sweetman, W L Nyhan
ABSTRACT
A double antibody technique has been developed to separate free biotin from bound biotin after competitive binding of [3H]biotin and unlabelled biotin to avidin. Antiavidin goat antibody was added followed by the addition of antigoat IgG antibody linked to agarose. Centrifugation separated the free biotin from the biotin bound to the avidin complex. The method was suitable for the detection of the amounts of biotin contained in 100-200 microliters of plasma or 5-10 microliters of urine. Normal values for the concentration of biotin in plasma and urine determined by this assay were 1.27 +/- 0.67 nmol/l and 49.1 +/- 35.7 mumol/mol creatinine, respectively.
Product Description
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Biotin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Biotin, meets USP testing specifications
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Sigma-Aldrich
Biotin, ≥99.0% (T)
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material