- Heteroarylureas with Spirocyclic Diamine Cores as Inhibitors of Fatty Acid Amide Hydrolase.
Heteroarylureas with Spirocyclic Diamine Cores as Inhibitors of Fatty Acid Amide Hydrolase.
Bioorganic & Medicinal Chemistry Letters (2014)
Keith JM, et al.