Merck
  • Abuse-related effects of µ-opioid analgesics in an assay of intracranial self-stimulation in rats: modulation by chronic morphine exposure.
Abuse-related effects of µ-opioid analgesics in an assay of intracranial self-stimulation in rats: modulation by chronic morphine exposure.
Behavioural pharmacology (2013-07-25)
Ahmad A Altarifi, Kenner C Rice, S Stevens Negus
ABSTRACT
Intracranial self-stimulation (ICSS) is an operant procedure in which responding is maintained by electrical brain stimulation. Stimulation frequency can be varied rapidly to maintain a wide range of baseline response rates, and drugs' effects can be evaluated simultaneously on both low ICSS rates maintained by low stimulation frequencies and high ICSS rates maintained by high stimulation frequencies. ICSS 'facilitation' indicates drug-induced increases in low ICSS rates and is often considered an abuse-related effect, whereas ICSS 'depression' indicates decreases in high ICSS rates and may indicate abuse-limiting effects. This study examined the roles of µ-agonist efficacy and of previous µ-agonist exposure as determinants of µ-agonist effects on ICSS in rats with electrodes implanted into the medial forebrain bundle. The high-efficacy, intermediate-efficacy, and low-efficacy µ agonists methadone, fentanyl, and nalbuphine were tested during escalating regimens of morphine exposure (vehicle, 3.2, and 18 mg/kg/day). During vehicle treatment, methadone and fentanyl primarily depressed ICSS, whereas nalbuphine produced weak facilitation that was not dose dependent. Chronic morphine produced tolerance to ICSS depression and increased expression of ICSS facilitation. These results suggest that µ-agonist exposure increases the expression of abuse-related ICSS facilitation by µ agonists with a broad range of efficacies at µ receptors.
Product Description
Supelco
Naltrexone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®