Merck
  • Normalyzer: a tool for rapid evaluation of normalization methods for omics data sets.
Normalyzer: a tool for rapid evaluation of normalization methods for omics data sets.
Journal of proteome research (2014-04-29)
Aakash Chawade, Erik Alexandersson, Fredrik Levander
ABSTRACT
High-throughput omics data often contain systematic biases introduced during various steps of sample processing and data generation. As the source of these biases is usually unknown, it is difficult to select an optimal normalization method for a given data set. To facilitate this process, we introduce the open-source tool "Normalyzer". It normalizes the data with 12 different normalization methods and generates a report with several quantitative and qualitative plots for comparative evaluation of different methods. The usefulness of Normalyzer is demonstrated with three different case studies from quantitative proteomics and transcriptomics. The results from these case studies show that the choice of normalization method strongly influences the outcome of downstream quantitative comparisons. Normalyzer is an R package and can be used locally or through the online implementation at http://quantitativeproteomics.org/normalyzer .
Sigma-Aldrich
Universal Proteomics Standard Set, Protein Mass Spectrometry Calibration Standard