Merck
  • Identification of bacteria pathogenic to or associated with onion (Allium cepa) based on sequence differences in a portion of the conserved gyrase B gene.
Identification of bacteria pathogenic to or associated with onion (Allium cepa) based on sequence differences in a portion of the conserved gyrase B gene.
Journal of microbiological methods (2014-06-14)
Jean M Bonasera, Jo Ann E Asselin, Steven V Beer
ABSTRACT
We have developed a method for the identification of Gram-negative bacteria, particularly members of the Enterobacteriaceae, based on sequence variation in a portion of the gyrB gene. Thus, we identified, in most cases to species level, over 1000 isolates from onion bulbs and leaves and soil in which onions were grown.
Sigma-Aldrich
Xylene Cyanol FF, BioReagent, Molecular Biology
Sigma-Aldrich
Xylene Cyanol FF, Dye content ≥75 %