Merck
  • Methods of multilocus enzyme electrophoresis for bacterial population genetics and systematics.
Methods of multilocus enzyme electrophoresis for bacterial population genetics and systematics.
Applied and environmental microbiology (1986-05-01)
R K Selander, D A Caugant, H Ochman, J M Musser, M N Gilmour, T S Whittam
Sigma-Aldrich
Citric acid monohydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, 99.5-102%
Sigma-Aldrich
Citric acid monohydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Citric acid monohydrate, reagent grade, ≥98% (GC/titration)