Merck
  • The pH-responsive behaviour of poly(acrylic acid) in aqueous solution is dependent on molar mass.
The pH-responsive behaviour of poly(acrylic acid) in aqueous solution is dependent on molar mass.
Soft matter (2016-01-30)
Thomas Swift, Linda Swanson, Mark Geoghegan, Stephen Rimmer
ABSTRACT
Fluorescence spectroscopy on a series of aqueous solutions of poly(acrylic acid) containing a luminescent label showed that polymers with molar mass, Mn < 16.5 kDa did not exhibit a pH responsive conformational change, which is typical of higher molar mass poly(acrylic acid). Below this molar mass, polymers remained in an extended conformation, regardless of pH. Above this molar mass, a pH-dependent conformational change was observed. Diffusion-ordered nuclear magnetic resonance spectroscopy confirmed that low molar mass polymers did not undergo a conformational transition, although large molar mass polymers did exhibit pH-dependent diffusion.
Sigma-Aldrich
2-Cyano-2-propyl benzodithioate, >97% (HPLC)
Pricing and availability is not currently available.