Merck
  • Improving the Performance of Organic Thin-Film Transistors by Ion Doping of Ethylene-Glycol-Based Self-Assembled Monolayer Hybrid Dielectrics.
Improving the Performance of Organic Thin-Film Transistors by Ion Doping of Ethylene-Glycol-Based Self-Assembled Monolayer Hybrid Dielectrics.
Advanced materials (Deerfield Beach, Fla.) (2015-11-03)
Hanno Dietrich, Simon Scheiner, Luis Portilla, Dirk Zahn, Marcus Halik
ABSTRACT
Tuning the electrostatics of ethylene-glycol-based self-assembled monolayers (SAMs) by doping with ions is shown. Molecular dynamics simulations unravel binding mechanisms and predict dipole strengths of the doped layers. Additionally, by applying such layers as dielectrics in organic thin-film transistors, the incorporated ions are proven to enhance device performance by lowering the threshold voltage and increasing conductivity.