Merck
  • Neuroprotection and CD131/GDNF/AKT Pathway of Carbamylated Erythropoietin in Hypoxic Neurons.
Neuroprotection and CD131/GDNF/AKT Pathway of Carbamylated Erythropoietin in Hypoxic Neurons.
Molecular neurobiology (2016-08-20)
Jing Ding, Jing Wang, Qin-Ying Li, Jie-Zhong Yu, Cun-Gen Ma, Xin Wang, Chuan-Zhen Lu, Bao-Guo Xiao
ABSTRACT
Carbamylated erythropoietin (CEPO), an EPO derivative, is attracting widespread interest due to neuroprotective effects without erythropoiesis. However, little is known about molecular mechanisms behind CEPO-mediated neuroprotection. In primary neurons with oxygen-glucose deprivation (OGD) and mice with hypoxia-reoxygenation, the neuroprotection and possible molecular mechanism of CEPO were performed by immunohistochemistry and immunocytochemistry, Western blot, RT-PCR, and ELISA. The comparisons were analyzed by ANOVA followed by unpaired two-tailed Student's t test. Both CEPO and EPO showed the neuroprotective effects in OGD model and hypoxic brain. CEPO did not trigger JAK-2 but activated AKT through glial cell line-derived neurotrophic factor (GDNF). It has been shown that CEPO acts upon a heteroreceptor complex comprising both the EPO receptor and the common β receptor subunit (βcR, also known as CD131). The blockage of CD131 reduced CEPO-mediated GDNF production, while GFR receptor blockage and GDNF neutralization inhibited CEPO-induced neurogenesis. Addition of GDNF to cultured neurons increased phosphorylation of AKT. CEPO protects neurons possible through the CD131/GDNF/AKT pathway.
Sigma-Aldrich
D-Lysine, ≥98% (HPLC)