Merck
  • HIV-1 enhances mTORC1 activity and repositions lysosomes to the periphery by co-opting Rag GTPases.
HIV-1 enhances mTORC1 activity and repositions lysosomes to the periphery by co-opting Rag GTPases.
Scientific reports (2017-07-16)
Alessandro Cinti, Valerie Le Sage, Miroslav P Milev, Fernando Valiente-Echeverría, Christina Crossie, Marie-Joelle Miron, Nelly Panté, Martin Olivier, Andrew J Mouland
ABSTRACT
HIV-1 co-opts several host machinery to generate a permissive environment for viral replication and transmission. In this work we reveal how HIV-1 impacts the host translation and intracellular vesicular trafficking machineries for protein synthesis and to impede the physiological late endosome/lysosome (LEL) trafficking in stressful conditions. First, HIV-1 enhances the activity of the master regulator of protein synthesis, the mammalian target of rapamycin (mTOR). Second, the virus commandeers mTOR-associated late endosome/lysosome (LEL) trafficking and counteracts metabolic and environmental stress-induced intracellular repositioning of LEL. We then show that the small Rag GTPases, RagA and RagB, are required for the HIV-1-mediated LEL repositioning that is likely mediated by interactions between the Rags and the viral proteins, Gag and Vif. siRNA-mediated depletion of RagA and RagB leads to a loss in mTOR association to LEL and to a blockade of viral particle assembly and release at the plasma membrane with a marked concomitant reduction in virus production. These results show that HIV-1 co-opts fundamental mechanisms that regulate LEL motility and positioning and support the notion that LEL positioning is critical for HIV-1 replication.
Sigma-Aldrich
MISSION® esiRNA, targeting human RRAGA
Sigma-Aldrich
5,6-Dichlorobenzimidazole 1-β-D-ribofuranoside
Sigma-Aldrich
Anti-Puromycin Antibody, clone 12D10, clone 12D10, from mouse