Merck
  • Membrane transporters and cytoplasmatic pH regulation on bovine Sertoli cells.
Membrane transporters and cytoplasmatic pH regulation on bovine Sertoli cells.
The Journal of membrane biology (2008-12-04)
P F Oliveira, M Sousa, A Barros, T Moura, A Rebelo da Costa
ABSTRACT
Sertoli cells are responsible for regulating a wide range of processes that lead to the differentiation of male germ cells into spermatozoa. Cytoplasmic pH (pHi) has been shown to be an important parameter in cell physiology, regulating namely cell metabolism and differentiation. However, membrane transport mechanisms involved in pHi regulation mechanisms of Sertoli cells have not yet been elucidated. In this work, pHi was determined using the pH-sensitive fluorescent probe 2',7'-bis-(2-carboxyethyl)-5-(and-6)-carboxyfluorescein (BCECF). Addition of weak acids resulted in rapid acidification of the intracellular milieu. Sertoli cells then recovered pHi by a mechanism that was shown to be sensitive to external Na+. pHi recovery was also greatly reduced in the presence of 4,4'-diisothiocyanatostilbene-2,2'-disulfonic acid (DIDS) and amiloride. These results point toward the action of an Na+-driven HCO3-/Cl- exchanger and/or an Na+/HCO3- cotransporter and the action of the Na+/H+ exchanger on pHi regulation in the experimental conditions used. pHi recovery was only slightly affected by ouabain, suggesting that the inhibition of Na+/K+-ATPase affects recovery indirectly, possibly via the shift on the Na+ gradient. On the other hand, recovery from the acid load was independent of the presence of concanamycin A, a specific inhibitor of the V-type ATPases, suggesting that these pumps do not have a relevant action on pHi regulation in bovine Sertoli cells.
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Standardized vial containing 2,000 Kunitz units of DNase I (D4527), vial of ≥0.25 mg total protein
Pricing and availability is not currently available.