Merck
  • Assessment practices of Irish speech and language therapists in the evaluation of voice disorders.
Assessment practices of Irish speech and language therapists in the evaluation of voice disorders.
Logopedics, phoniatrics, vocology (2017-01-05)
Ciarán Kenny
ABSTRACT
It is commonly accepted that the evaluation of voice disorders ought to include extensive perceptual, psychometric, and instrumental measurements. This serves to encapsulate the wide-reaching effects of such a disorder, from the physical impairment in voice production to the psycho-social impact of having a dysphonic voice. In spite of this, no international gold standard exists by which voice disorders should be evaluated, and so speech and language therapists (SLTs) are often tasked with developing an assessment battery for use in their own clinics. The purpose of this study is to determine whether the evaluation of voice disorders by Irish SLTs on a national scale is suitably comprehensive, with particular reference to the guidelines published by the European Laryngological Society. A total of 49 SLTs working in a variety of settings responded anonymously to an electronic survey regarding their assessment practices. Results indicate that therapists are comprehensive in non-instrumental evaluation of voice, but lack both access to and training in instrumental assessment techniques.
Product Description
Supelco
VOCOL® Capillary GC Column, L × I.D. 60 m × 0.25 mm, df 1.50 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 3.00 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 1.80 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 1.50 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 105 m × 0.53 mm, df 3.00 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 3.00 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 60 m × 0.53 mm, df 3.00 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 20 m × 0.18 mm, df 1.00 μm