Merck
  • Silenced suppressor of cytokine signaling 1 (SOCS1) enhances the maturation and antifungal immunity of dendritic cells in response to Candida albicans in vitro.
Silenced suppressor of cytokine signaling 1 (SOCS1) enhances the maturation and antifungal immunity of dendritic cells in response to Candida albicans in vitro.
Immunologic research (2014-11-10)
Dongmei Shi, Dongmei Li, Qingxin Yin, Ying Qiu, Hongxia Yan, Yongnian Shen, Guixia Lu, Weida Liu
ABSTRACT
Dendritic cells (DCs) are known to play an important role in initiating and orchestrating antimicrobial immunity. Given the fact that candidiasis appears often in immunocompromised patients, it seems plausible that DCs hold the key to new antifungal strategies. One possibility to enhance the potency of DC-based immunotherapy is to silence the negative immunoregulatory pathways through the ablation suppressor of cytokine signaling suppressor 1 (SOCS1). Here, we deliver small interfering RNA (siRNA) against SOCS1 into murine bone marrow DCs, and as a consequence, we investigate the maturation/action of DCs and the subsequent T cell response after exposure to C. albicans. Our results show that the maturation of DCs (i.e., expressions of CD80, CD40, CD86, and MHC II) are significantly increased in the silenced SOCS1 treatment group after exposure to C. albicans. As a result, suppression of the SOCS1 promotes the greatest expression of IFN-γ and IL-12, and reduces IL-4 secretions, which induce CD4(+) cell Th1 differentiation but inactivate Th2 cell development. The responses of IL-6 and TNF-β consist of up-regulation in the presence of C. albicans, but this is not specific to SOCS1 silencing, suggesting that these cytokines are not regulated by the SOCS1 gene in fungal infections. We find Th17 differentiation is unchanged regardless of SOCS1 inhibition. The increase in phagocytosis and killing of C. albicans in SOCS1 gene-treated DCs indicate a role for this cytokine suppressor in innate immunity as well. In conclusion, our findings support the view that SOCS1 protein is a critical inhibitory molecule for controlling cytokine response and antigen presentation by DCs, thereby regulating the magnitude of innate and adaptive immunities by generating IFN-γ-production T cells (Th1)--but not Th17--from naïve CD4(+) T cells. Our study demonstrates that SOCS1 siRNA can serve as a useful vehicle to modulate the function of DCs against C. albicans infection.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Bicarbonato di sodio, ACS reagent, ≥99.7%
Sigma-Aldrich
Bicarbonato di sodio, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Bicarbonato di sodio, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Trypan Blue, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Bicarbonato di sodio, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Brefeldina A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Trypan Blue, 0.4%, suitable for microscopy
Sigma-Aldrich
Brefeldina A, ≥99% (HPLC and TLC), BioXtra, Molecular Biology
Sigma-Aldrich
Bicarbonato di sodio, BioXtra, 99.5-100.5%
USP
Bicarbonato di sodio, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Brefeldina A, Ready Made Solution, from Penicillium brefeldianum, 10 mg/mL in DMSO
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Bicarbonato di sodio, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Bicarbonato di sodio, −40-+140 mesh, ≥95%
Sigma-Aldrich
Bicarbonato di sodio, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, suitable for fluorescence, ≥95.0% (HPLC)