Merck
  • Development of a morphogenetically active scaffold for three-dimensional growth of bone cells: biosilica-alginate hydrogel for SaOS-2 cell cultivation.
Development of a morphogenetically active scaffold for three-dimensional growth of bone cells: biosilica-alginate hydrogel for SaOS-2 cell cultivation.
Journal of tissue engineering and regenerative medicine (2013-04-16)
Werner E G Müller, Heinz C Schröder, Qingling Feng, Ute Schlossmacher, Thorben Link, Xiaohong Wang
ABSTRACT
Polymeric silica is formed from ortho-silicate during a sol-gel formation process, while biosilica is the product of an enzymatically driven bio-polycondensation reaction. Both polymers have recently been described as a template that induces an increased expression of the genes encoding bone morphogenetic protein 2 (BMP-2) and osteoprotegerin in osteoblast-related SaOS-2 cells; simultaneously or subsequently the cells respond with enhanced hydroxyapatite formation. In order to assess whether the biocompatible polymeric silica/biosilica can serve as a morphogenetically active matrix suitable for three-dimensional (3D) cell growth, or even for 3D cell bioprinting, SaOS-2 cells were embedded into a Na-alginate-based hydrogel. Four different gelatinous hydrogel matrices were used for suspending SaOS-2 cells: (a) the hydrogel alone; (b) the hydrogel with 400 μM ortho-silicate; (c) the hydrogel supplemented with 400 μM ortho-silicate and recombinant silicatein to allow biosilica synthesis to occur; and (d) the hydrogel with ortho-silicate and BSA. The SaOS-2 cells showed an increased growth if silica/biosilica components were present in the hydrogel. Likewise intensified was the formation of hydroxyapatite nodules in the silica-containing hydrogels. After an incubation period of 2 weeks, cells present in silica-containing hydrogels showed a significantly higher expression of the genes encoding the cytokine BMP-2, the major fibrillar structural protein collagen 1 and likewise of carbonic anhydrase. It is concluded that silica, and to a larger extent biosilica, retains its morphogenetic/osteogenic potential after addition to Na-alginate-based hydrogels. This property might qualify silica hydrogels to be also used as a matrix for 3D cell printing.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
tetra-etil ortosilicato, reagent grade, 98%
Sigma-Aldrich
Desametasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Acido L-ascorbico, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Acido L-ascorbico, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
tetra-etil ortosilicato, ≥99.0% (GC)
Sigma-Aldrich
Acido L-ascorbico, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Acido L-ascorbico, 99%
Sigma-Aldrich
Acido L-ascorbico, reagent grade, crystalline
Sigma-Aldrich
Desametasone, ≥98% (HPLC), powder
Sigma-Aldrich
Acido L-ascorbico, ACS reagent, ≥99%
Supelco
Acido L-ascorbico, analytical standard
Sigma-Aldrich
tetra-etil ortosilicato, 99.999% trace metals basis
Sigma-Aldrich
Acido L-ascorbico, meets USP testing specifications
Sigma-Aldrich
Acido L-ascorbico, reagent grade
Sigma-Aldrich
Desametasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Acido L-ascorbico, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Acido L-ascorbico, FCC, FG
Sigma-Aldrich
Acido L-ascorbico, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Acido L-ascorbico, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Desametasone, meets USP testing specifications
Sigma-Aldrich
tetra-etil ortosilicato, packaged for use in deposition systems
Sigma-Aldrich
Thrombopoietin human, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥98% (SDS-PAGE and HPLC)
Sigma-Aldrich
Thrombopoietin from mouse, recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture, >97% (SDS-PAGE)