Merck
  • Identification of potential HIV restriction factors by combining evolutionary genomic signatures with functional analyses.
Identification of potential HIV restriction factors by combining evolutionary genomic signatures with functional analyses.
Retrovirology (2015-05-20)
Paul J McLaren, Ali Gawanbacht, Nitisha Pyndiah, Christian Krapp, Dominik Hotter, Silvia F Kluge, Nicola Götz, Jessica Heilmann, Katharina Mack, Daniel Sauter, Danielle Thompson, Jérémie Perreaud, Antonio Rausell, Miguel Munoz, Angela Ciuffi, Frank Kirchhoff, Amalio Telenti
ABSTRACT
Known antiretroviral restriction factors are encoded by genes that are under positive selection pressure, induced during HIV-1 infection, up-regulated by interferons, and/or interact with viral proteins. To identify potential novel restriction factors, we performed genome-wide scans for human genes sharing molecular and evolutionary signatures of known restriction factors and tested the anti-HIV-1 activity of the most promising candidates. Our analyses identified 30 human genes that share characteristics of known restriction factors. Functional analyses of 27 of these candidates showed that over-expression of a strikingly high proportion of them significantly inhibited HIV-1 without causing cytotoxic effects. Five factors (APOL1, APOL6, CD164, TNFRSF10A, TNFRSF10D) suppressed infectious HIV-1 production in transfected 293T cells by >90% and six additional candidates (FCGR3A, CD3E, OAS1, GBP5, SPN, IFI16) achieved this when the virus was lacking intact accessory vpr, vpu and nef genes. Unexpectedly, over-expression of two factors (IL1A, SP110) significantly increased infectious HIV-1 production. Mechanistic studies suggest that the newly identified potential restriction factors act at different steps of the viral replication cycle, including proviral transcription and production of viral proteins. Finally, we confirmed that mRNA expression of most of these candidate restriction factors in primary CD4+ T cells is significantly increased by type I interferons. A limited number of human genes share multiple characteristics of genes encoding for known restriction factors. Most of them display anti-retroviral activity in transient transfection assays and are expressed in primary CD4+ T cells.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Acido solforico, 99.999%
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin tested
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Interleukin-2, human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Interleukin-2 from mouse, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
IL-2 from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Interleukin-2 from mouse, IL-2, recombinant, expressed in E. coli, carrier free
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in E. coli, ~10000 U/mL
Prezzi e disponibilità al momento non sono disponibili