表示を変更する
| サイズ/SKU | 在庫状況 | 単価 |
|---|---|---|
1 ea | カートの在庫状況を確認する | ¥18,300 |
この商品について
NACRES:
NB.51
UNSPSC Code:
41121809
Joint:
Joints: (27)
Packaging:
pkg of 1 ea
¥18,300
カートの在庫状況を確認する
joint
Joints: (27)
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F477500A
capacity
500 mL , 500 mL
General description
Supplied with a Glass or PTFE stopper
Legal Information
Synthware is a trademark of Kemtech America Inc.
1 of 1
当該品目 | |||
|---|---|---|---|
| description 500 mL, Joints: (27) | description 1000 mL, Joints: (27) | description 500 mL, Joints: (27) | description 1000 mL, Joints: (27) |
| manufacturer/tradename Synthware F477500A | manufacturer/tradename Synthware F477000A | manufacturer/tradename Synthware F477500 | manufacturer/tradename Synthware F477000 |
| capacity 500 mL, 500 mL | capacity 1000 mL, 1000 mL | capacity 500 mL , 500 mL | capacity 1000 mL , 1000 mL |
| joint Joints: (27) | joint Joints: (27) | joint Joints: (27) | joint Joints: (27) |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
Still not finding the right product?
Explore all of our products under Synthware™ Squibb-style separatory funnel with PTFE stopcock and Glass stopper