Merck
  • Analysis of phenolics in wine by high performance thin-layer chromatography with gradient elution and high resolution plate imaging.
Analysis of phenolics in wine by high performance thin-layer chromatography with gradient elution and high resolution plate imaging.
Journal of pharmaceutical and biomedical analysis (2014-09-26)
Snezana Agatonovic-Kustrin, Chandima G Hettiarachchi, David W Morton, Slavica Razic
RESUMEN
Health benefits of wine, especially with red wine, have been linked to the presence of a wide range of phenolic antioxidants. Thus, the aim of this study was to develop a simple, high performance thin layer chromatographic (HPTLC) method combined with high resolution digital plate images to visually compare multiple wine samples simultaneously on a single chromatographic plate and to quantify levels of gallic acid, caffeic acid, resveratrol and rutin, as representatives of the four different classes of phenolics found in wines. We also wanted to investigate the contribution of the investigated phenolic compounds to the total polyphenolic content (TPC) and total antioxidant capacity (TAC) of the wine samples. The average concentrations of caffeic acid, gallic acid, resveratrol, and rutin in the red wines were 2.15, 30.17, 0.59 and 2.47 mg/L respectively with their concentration below limit of quantification in the white wine samples. The highest concentration of resveratrol and rutin is found in the Cabernet and Shiraz wine samples. The amounts of gallic acid are correlated with TPC (r=0.58). Italian wines have the highest correlation between TPC and TAC (r=0.99) although they do not contain detectable amounts of resveratrol, they contain significant amount of rutin. Therefore, antioxidant properties might be associated with the presence of flavanols in these wines.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Metanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Diclorometano, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Metanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Heptano, suitable for HPLC, ≥99%
Sigma-Aldrich
Diclorometano, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Ácido fórmico, reagent grade, ≥95%
Sigma-Aldrich
Diclorometano, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Metanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Dodecilsulfatosódico, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Diclorometano, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Heptano, ReagentPlus®, 99%
Sigma-Aldrich
Ácido fórmico, ACS reagent, ≥96%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
Ácido fórmico, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Metanol, anhydrous, 99.8%
Sigma-Aldrich
Dodecilsulfatosódico, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
Dodecilsulfatosódico, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Ácido fórmico, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Metanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Metanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Metanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Dodecilsulfatosódico, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Diclorometano, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Heptano, HPLC Plus, for HPLC, GC, and residue analysis, 99%
Sigma-Aldrich
Ácido fórmico, ACS reagent, ≥88%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Heptano, puriss. p.a., reag. Ph. Eur., ≥99% n-heptane basis (GC)