Przejdź do
Inchi Code
1S/C88H128N12O19/c1-15-59(8)79(71(113-13)53-75(104)99-44-24-32-70(99)81(114-14)60(9)82(106)92-61(10)80(105)65-27-17-16-18-28-65)97(11)86(110)77(57(4)5)96-85(109)78(58(6)7)98(12)88(112)119-55-62-33-37-67(38-34-62)93-83(107)68(30-23-42-91-87(89)111)94-84(108)76(56(2)3)95-73(102)41-45-115-47-49-117-51-52-118-50-48-116-46-43-90-72(101)39-40-74(103)100-54-66-29-20-19-25-63(66)35-36-64-26-21-22-31-69(64)100/h16-22,25-29,31,33-34,37-38,56-61,68,70-71,76-81,105H,15,23-24,30,32,39-55H2,1-14H3,(H,90,101)(H,92,106)(H,93,107)(H,94,108)(H,95,102)(H,96,109)(H3,89,91,111)/t59-,60+,61+,68-,70-,71+,76-,77-,78-,79-,80+,81+/m0/s1
Inchi Key
LJJUBMUNOGZMDD-UDEUCFDSSA-N
Shipping Temperature
Cold-chain transportation
Purity
95%
Storage temp.
Inert atmosphere, store in freezer, under -20C
Country of Origin
CN
Physical Form
White to yellow powder or crystals
hstatements
signalword
Warning
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?