Przejdź do zawartości
Merck

Przejdź do

C2160 Human Blood, ECACC 950515239

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Zaloguj się, aby wyświetlić ceny organizacyjne i kontraktowe.

Wybierz wielkość

Zmień widok
Rozmiar/SKUDostępnośćCena netto

Informacje o tej pozycji

UNSPSC Code:
41106514
Biological source:
human blood
Technique(s):
cell culture | mammalian: suitable
Ethnicity:
UK Caucasian
Gender:
male
Growth mode:
Suspension
Morphology:
Lymphoblastoid
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Potrzebujesz pomocy? Nasz zespół doświadczonych naukowców chętnie Ci pomoże.
Pozwól nam pomóc


Nazwa produktu

C2160, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

biological source

human blood

manufacturer/tradename

ECACC

packaging

tube of 5 μg 950515239-DNA-5UG, pkg of vial of cells 950515239-1VL

HRC Panel Number

4

ethnicity

UK Caucasian

gender

male

growth mode

Suspension

karyotype

Not Specified

morphology

Lymphoblastoid

technique(s)

cell culture | mammalian: suitable

Application

As part of the Human Random Control collection this cell line can be used as a renewable source of genomic DNA reference material for a wide range of genetic research applications.

Biochem/physiol Actions

A B-lymphoblastoid cell line derived from a randomly selected Caucasian blood donor whose parents and grandparents were born in the UK or Ireland. The cell line was generated by Epstein-Barr Virus (EBV) transformation of peripheral blood lymphocytes.

This cell line is part of the European Collection of Cell Cultures (ECACC) Human Random Control (HRC) collection. The HRC collection consists of more than 700 B-lymphoblastoid cell lines derived from randomly selected Caucasian blood donors whose parents and grandparents were born in the UK or Ireland. The HRC collection can be used as a renewable control population for genetic research.
UK Caucasian Male

Preparation Note

RPMI 1640 (R0883) + 2 mM L-Glutamine (G7513) + 20% FBS / FCS (F2442). Once a growing culture is established the FBS concentration can be reduced to 10%.
Maintain cultures between 3 x 105 - 2 x 106 cells/ml; 5% CO2, 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Ta strona może zawierać tekst przetłumaczony maszynowo.

Porównaj podobne pozycje

Wyświetl pełne porównanie

Pokaż różnice

1 of 1

Ta pozycja
CB_95051589CB_950515191CB_950515194
vibrant-m

C2160

vibrant-m

C2102

vibrant-m

C2143

vibrant-m

C2146

description

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

description

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

description

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

description

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

morphology

Lymphoblastoid

morphology

Lymphoblastoid

morphology

Lymphoblastoid

morphology

Lymphoblastoid

packaging

tube of 5 μg 950515239-DNA-5UG, pkg of vial of cells 950515239-1VL

packaging

pkg of vial of cells 95051589-1VL, tube of 5 μg 95051589-DNA-5UG

packaging

-

packaging

tube of 5 μg 950515194-DNA-5UG, pkg of vial of cells 950515194-1VL

ethnicity

UK Caucasian

ethnicity

UK Caucasian

ethnicity

UK Caucasian

ethnicity

UK Caucasian

manufacturer/tradename

ECACC

manufacturer/tradename

ECACC

manufacturer/tradename

ECACC

manufacturer/tradename

ECACC

growth mode

Suspension

growth mode

Suspension

growth mode

-

growth mode

Suspension

biological source

human blood

biological source

human blood

biological source

-

biological source

human blood




Wybierz jedną z najnowszych wersji:

Certyfikaty analizy (CoA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumenty section.

Proszę o kontakt, jeśli potrzebna jest pomoc Obsługa Klienta

Masz już ten produkt?

Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.

Odwiedź Bibliotekę dokumentów



Questions

Reviews

No rating value

Active Filters