Przejdź do
Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
Nazwa produktu
C2102, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
biological source
human blood
manufacturer/tradename
ECACC
packaging
tube of 5 μg 95051589-DNA-5UG, pkg of vial of cells 95051589-1VL
HRC Panel Number
4
ethnicity
UK Caucasian
gender
male
growth mode
Suspension
karyotype
Not Specified
morphology
Lymphoblastoid
technique(s)
cell culture | mammalian: suitable
Application
Biochem/physiol Actions
This cell line is part of the European Collection of Cell Cultures (ECACC) Human Random Control (HRC) collection. The HRC collection consists of more than 700 B-lymphoblastoid cell lines derived from randomly selected Caucasian blood donors whose parents and grandparents were born in the UK or Ireland. The HRC collection can be used as a renewable control population for genetic research.
Preparation Note
Other Notes
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA. | description NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA. | description NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA. | description NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA. |
| growth mode Suspension | growth mode Suspension | growth mode Suspension | growth mode Suspension |
| ethnicity UK Caucasian | ethnicity UK Caucasian | ethnicity UK Caucasian | ethnicity - |
| manufacturer/tradename ECACC | manufacturer/tradename ECACC | manufacturer/tradename ECACC | manufacturer/tradename ECACC |
| biological source human blood | biological source human blood | biological source - | biological source human blood |
| morphology Lymphoblastoid | morphology Lymphoblastoid | morphology Lymphoblastoid | morphology - |
| packaging pkg of vial of cells 95051589-1VL, tube of 5 μg 95051589-DNA-5UG | packaging pkg of vial of cells 95061993-1VL, tube of 5 μg 95061993-DNA-5UG | packaging - | packaging - |
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?
Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.
