Merck
Acid Suspends the Circadian Clock in Hypoxia through Inhibition of mTOR.
Cell (2018-06-05)
Zandra E Walton, Chirag H Patel, Rebekah C Brooks, Yongjun Yu, Arig Ibrahim-Hashim, Malini Riddle, Alessandra Porcu, Tianying Jiang, Brett L Ecker, Feven Tameire, Constantinos Koumenis, Ashani T Weeraratna, David K Welsh, Robert Gillies, James C Alwine, Lin Zhang, Jonathan D Powell, Chi V Dang
ABSTRAKT
Recent reports indicate that hypoxia influences the circadian clock through the transcriptional activities of hypoxia-inducible factors (HIFs) at clock genes. Unexpectedly, we uncover a profound disruption of the circadian clock and diurnal transcriptome when hypoxic cells are permitted to acidify to recapitulate the tumor microenvironment. Buffering against acidification or inhibiting lactic acid production fully rescues circadian oscillation. Acidification of several human and murine cell lines, as well as primary murine T cells, suppresses mechanistic target of rapamycin complex 1 (mTORC1) signaling, a key regulator of translation in response to metabolic status. We find that acid drives peripheral redistribution of normally perinuclear lysosomes away from perinuclear RHEB, thereby inhibiting the activity of lysosome-bound mTOR. Restoring mTORC1 signaling and the translation it governs rescues clock oscillation. Our findings thus reveal a model in which acid produced during the cellular metabolic response to hypoxia suppresses the circadian clock through diminished translation of clock constituents.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Goat serum
Sigma-Aldrich
Triton X-100, laboratory grade
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Triton X-100, Molecular Biology
Sigma-Aldrich
DMOG, ≥98% (HPLC)
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without glucose and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Torin2, ≥98% (HPLC)
Sigma-Aldrich
Amiloride hydrochloride hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)