Merck
Human Mps1 protein kinase is required for centrosome duplication and normal mitotic progression.
Proceedings of the National Academy of Sciences of the United States of America (2003-12-06)
Harold A Fisk, Christopher P Mattison, Mark Winey
ABSTRAKT
The mitotic spindle is essential for the maintenance of genetic stability, and in budding yeast its assembly and function depend on the Mps1 protein kinase. Mps1p is required for centrosome duplication and the spindle checkpoint. Several recent reports demonstrate that vertebrate Mps1 proteins regulate the spindle checkpoint, but reports conflict regarding their role in centrosome duplication. Here we provide multiple lines of evidence that the human Mps1 protein (hMps1) is required for centrosome duplication. A recently described rabbit polyclonal antibody against hMps1 specifically recognizes centrosomes in a variety of human cell types. Overexpression of a dominant-negative version of hMps1 (hMps1KD) can prevent centrosome duplication in a variety of cell types, and active hMps1 accelerates centrosome reduplication in U2OS cells. Finally, we demonstrate that disruption of hMps1 function with pools of hMps1-specific small interfering RNAs causes a pleiotropic phenotype resulting from the combination of severe mitotic abnormalities and failures in centrosome duplication. This approach demonstrates that hMps1 is required for centrosome duplication and for the normal progression of mitosis, and suggests that the threshold level of hMps1 function required for centrosome duplication is lower than that required for hMps1 mitotic functions.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Informacje o cenach i dostępności nie są obecnie dostępne.