Merck
Insights into the hierarchical structure and digestibility of starch in heat-moisture treated adlay seeds.
Food chemistry (2020-03-07)
Hongwei Wang, Jiangtao Ding, Naiyong Xiao, Xingli Liu, Yanyan Zhang, Hua Zhang
ABSTRAKT
In the present study, starches isolated from heat-moisture treated (HMT) adlay seeds were characterized with hierarchical structures and digestibility to understand the relationships in structural and digestible behaviors of starches in a HMT starch-based food system. The results indicated that HMT not only caused the disorganizations of hierarchical structures, but also promoted the molecular rearrangements and reassembly of starch chains to form a new ordered molecular aggregation architecture (i.e., highly ordered molecular structure, amylose-lipids complexes and thicker crystalline lamella). And the reinforced molecular interactions between starch molecules during HMT occurred to form more densely ordered structure, thus reducing the digestibility and digestion rate coefficients (k) of adlay starch. Particularly, HMT applied directly in adlay seeds with 30% moisture content displayed the highest resistant starch level (20.6%) and lowest k (2.74 × 10-3 min-1). Thus, the results are useful to realize the desired regulation of starch digestibility in adlay products following HMT.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pancreatin from porcine pancreas, 8 × USP specifications
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Amylopectin from potato starch
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Amylose from potato, used as amylase substrate
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
α-Amylase, heat-stable, solution, for use in Total Dietary Fiber Assay, TDF-100A
Informacje o cenach i dostępności nie są obecnie dostępne.