Merck
PRC2 Acts as a Critical Timer That Drives Oligodendrocyte Fate over Astrocyte Identity by Repressing the Notch Pathway.
Cell reports (2020-09-17)
Wenxian Wang, Hyeyoung Cho, Dongkyeong Kim, Younjung Park, Ji Hwan Moon, Su Jeong Lim, Sung Min Yoon, Michael McCane, Sue A Aicher, Sangsoo Kim, Ben Emery, Jae W Lee, Seunghee Lee, Yungki Park, Soo-Kyung Lee
ABSTRAKT
PRC2 creates the repressive mark histone H3 Lys27 trimethylation. Although PRC2 is involved in various biological processes, its role in glial development remains ambiguous. Here, we show that PRC2 is required for oligodendrocyte (OL) differentiation and myelination, but not for OL precursor formation. PRC2-deficient OL lineage cells differentiate into OL precursors, but they fail to trigger the molecular program for myelination, highlighting that PRC2 is essential for directing the differentiation timing of OL precursors. PRC2 null OL lineage cells aberrantly induce Notch pathway genes and acquire astrocytic features. The repression of the Notch pathway restores the myelination program and inhibits abnormal astrocytic differentiation in the PRC2-deficient OL lineage, indicating that Notch is a major target of PRC2. Altogether, our studies propose a specific action of PRC2 as a novel gatekeeper that determines the glial fate choice and the timing of OL lineage progression and myelination by impinging on the Notch pathway.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Przeciwciało przeciw trimetylo-histonowi H3 (Lys27), Upstate®, from rabbit
Sigma-Aldrich
Anti-Galactocerebroside Antibody, clone mGalC, clone mGalC, Chemicon®, from mouse
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, a.a. 82-87, culture supernatant, clone 12, Chemicon®
Sigma-Aldrich
ChIPAb+ EED - przeciwciało i zestaw primerów zweryfikowane pod kątem ChIP, from mouse
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
XAV939, ≥98% (HPLC)
Sigma-Aldrich
Anti-CNPase Antibody, clone 11-5B, clone 11-5B, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, ascites fluid, clone GA5, Chemicon®
Sigma-Aldrich
LY-411575, ≥97% (HPLC)