Merck
  • Anti-inflammatory activity of verminoside from Kigelia africana and evaluation of cutaneous irritation in cell cultures and reconstituted human epidermis.
Anti-inflammatory activity of verminoside from Kigelia africana and evaluation of cutaneous irritation in cell cultures and reconstituted human epidermis.
Journal of natural products (2005-11-29)
Patrizia Picerno, Giuseppina Autore, Stefania Marzocco, Marisa Meloni, Rokia Sanogo, Rita P Aquino
ABSTRAKT
Kigelia africana is a plant used in Africa for anti-inflammatory, anti-microbial, and anti-skin-aging effects. Various papers have reported on the composition and biological activities of its CH2Cl2 extracts and dermal formulations. Chemical analysis of a polar extract of fruit from K. africana indicated the presence of verminoside (1), an iridoid, as a major constituent, and of a series of polyphenols such as verbascoside (2). In vitro assays showed that 1 had significant anti-inflammatory effects, inhibiting both iNOS expression and NO release in the LPS-induced J774.A1 macrophage cell line. Cytotoxicity and cutaneous irritation of the extract and of compounds 1 and 2 were investigated. The crude extract and 1 did not affect cell viability in vitro either in cells grown in monolayers (ML) or in the reconstituted human epidermis (RHE, 3D) model; neither caused release of pro-inflammatory mediators or histomorphological modification of RHE.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri