- Isoechinulin-type alkaloids, variecolorins A-L, from halotolerant Aspergillus variecolor.
Isoechinulin-type alkaloids, variecolorins A-L, from halotolerant Aspergillus variecolor.
Journal of natural products (2007-09-28)
Wen-Liang Wang, Zhen-Yu Lu, Hong-Wen Tao, Tian-Jiao Zhu, Yu-Chun Fang, Qian-Qun Gu, Wei-Ming Zhu
PMID17896816
ABSTRAKT
Twelve new compounds, variecolorins A-L ( 1- 12), together with eleven known analogues ( 13- 23) were isolated from the broth of a halotolerant fungus, Aspergillus variecolor. The structures of compounds 1- 12 were determined by chemical and spectroscopic methods. Compounds 1- 11, 13- 15, and 20- 23 exhibited weak radical scavenging activity against DPPH, with IC 50 values from 43 to 103 microM. The new compounds 1- 12 all were essentially nontoxic against the P388, HL-60, BEL-7402, and A-549 cell lines with IC 50 values from 70 to 260 microM.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)