Merck
Non-heme dioxygenases: cellular sensors and regulators jelly rolled into one?
Nature chemical biology (2007-02-16)
Abdullah Ozer, Richard K Bruick
ABSTRAKT
Members of the Fe(II)- and 2-oxoglutarate-dependent family of dioxygenases have long been known to oxidize several amino acids in various protein targets to facilitate protein folding. However, in recent years investigators have characterized several such hydroxylation modifications that serve a regulatory, rather than structural, purpose. Furthermore, the responsible enzymes seem to function directly as sensors of the cellular environment and metabolic state. For example, a cellular response pathway to low oxygen (hypoxia) is orchestrated through the actions of prolyl and asparaginyl hydroxylases that govern both the oxygen-dependent stability and transcriptional activity of the hypoxia-inducible transcription factor. Recently, a different subfamily of Fe(II)- and 2-oxoglutarate-dependent dioxygenases has been shown to carry out histone demethylation. The discovery of protein regulation via hydroxylation raises the possibility that other Fe(II)- and 2-oxoglutarate-dependent dioxygenases might also serve in a similar capacity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
α-Ketoglutaric acid, ≥98.5% (NaOH, titration)
Sigma-Aldrich
α-Ketoglutaric acid, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
α-Ketoglutaric acid, 99.0-101.0% (T)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)