Merck
Bioactivities of simplified adociaquinone B and naphthoquinone derivatives against Cdc25B, MKP-1, and MKP-3 phosphatases.
Bioorganic & medicinal chemistry (2008-11-26)
Shugeng Cao, Brian T Murphy, Caleb Foster, John S Lazo, David G I Kingston
ABSTRAKT
Some simplified adociaquinone B analogs and a series of 1,4-naphthoquinone derivatives were synthesized and tested against the three enzymes Cdc25B, MKP-1, and MKP-3. Cdc25B and MKP-1 in particular are enzymes overexpressed in human cancer cells, and they represent potential molecular targets for novel cancer chemotherapeutic treatments. A number of analogs exhibited significant inhibitory activity against these enzymes, and the bioassay data in addition to structure-activity relationships of these compounds will be discussed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Menadione (K3), analytical standard
Sigma-Aldrich
Menadione, crystalline
Sigma-Aldrich
Menadione, meets USP testing specifications
Sigma-Aldrich
5-Hydroxy-1,4-naphthoquinone, 97%
Sigma-Aldrich
5,8-Dihydroxy-1,4-naphthoquinone, technical grade
Sigma-Aldrich
1,4-Naphthoquinone, purum, ≥96.5% (HPLC)
Sigma-Aldrich
1,4-Naphthoquinone, 97%
Supelco
Melting point standard 283-286°C, analytical standard
Sigma-Aldrich
Anthraquinone, 97%
Sigma-Aldrich
p-Benzoquinone, reagent grade, ≥98%
Supelco
p-Benzoquinone, for spectrophotometric det. of amines, ≥99.5% (HPLC)