Merck
QSPR modeling of octanol/water partition coefficient of antineoplastic agents by balance of correlations.
European journal of medicinal chemistry (2010-01-29)
Andrey A Toropov, Alla P Toropova, Ivan Raska, Emilio Benfenati
ABSTRAKT
Three different splits into the subtraining set (n = 22), the set of calibration (n = 21), and the test set (n = 12) of 55 antineoplastic agents have been examined. By the correlation balance of SMILES-based optimal descriptors quite satisfactory models for the octanol/water partition coefficient have been obtained on all three splits. The correlation balance is the optimization of a one-variable model with a target function that provides both the maximal values of the correlation coefficient for the subtraining and calibration set and the minimum of the difference between the above-mentioned correlation coefficients. Thus, the calibration set is a preliminary test set.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)
Sigma-Aldrich
Chlorambucil
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
2-Fluoroadenine-9-β-D-arabinofuranoside, DNA synthesis and methylation inhibitor
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Supelco
2,4′-DDD, PESTANAL®, analytical standard
Sigma-Aldrich
Temozolomide, ≥98% (HPLC)
Sigma-Aldrich
6-Thioguanine, ≥98%
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, meets USP testing specifications
Supelco
Busulfan, analytical standard, for drug analysis
Supelco
Temozolomide, VETRANAL®, analytical standard
Sigma-Aldrich
Melphalan, powder