Merck
  • Facile synthesis and stereochemical investigation of Mannich base derivatives: evaluation of antioxidant property and antituberculostic potency.
Facile synthesis and stereochemical investigation of Mannich base derivatives: evaluation of antioxidant property and antituberculostic potency.
Bioorganic & medicinal chemistry letters (2011-03-25)
Paramasivam Parthiban, Viswalingam Subalakshmi, Krishnamurthy Balasubramanian, Md Nurul Islam, Jae Sue Choi, Yeon Tae Jeong
ABSTRAKT
A mini-library of diversely substituted 2,4-diaryl-3-azabicyco[3.3.1]nonan-9-one O-methyloximes and their N-methyl analogs were synthesized by a non-laborious, modified and an optimized Mannich condensation in good yields. Both the ring N-methylation and oxime O-methylation were employed by various methods; of them, the usage of (t)BuOK was found to be the superior in terms of good yield in short time. Stereochemistry of all the synthesized compounds was unambiguously established by their NMR spectral ((1)H, (13)C, (1)H-(1)H COSY, (1)H-(13)C one and multiple bond COSY and NOESY) as well as single-crystal XRD studies. Irrespective of the nature and position of the substituents, all the synthesized oxime ethers of the bicyclic Mannich bases as well as their N-methyl analogs adopted the twin-chair conformation with equatorial orientations of all the substituents. All the synthesized oxime ethers were evaluated for their antioxidant property by DPPH radical scavenging method. According to the structure-activity correlations, compound 4y was found to be a lead molecule with the IC(50) of 0.187 mg/mL. Thus, the present study exploits the scope of finding more active analogs by further optimization with the incorporation of more electron enriched alkoxy/amino and/or phenolic groups on the heterocycle as well as oxime ether pharmacophore. Most of the synthesized molecules were screened for their antituberculostic potency against Mycobacterium tuberculosis H(37)Rv by zone of inhibition method. Of them, 4w/5d and 4x showed very promising inhibition zones of 21 and 23 mm, respectively, which leads to the optimization of 4x by introducing various substituents on the O-benzyl moiety to enhance the antituberculostic potency.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Isoniazid, analytical standard, ≥99% (TLC)
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%