Merck
Genetic validation of PfFKBP35 as an antimalarial drug target.
eLife (2023-11-07)
Basil T Thommen, Jerzy M Dziekan, Fiona Achcar, Seth Tjia, Armin Passecker, Katarzyna Buczak, Christin Gumpp, Alexander Schmidt, Matthias Rottmann, Christof Grüring, Matthias Marti, Zbynek Bozdech, Nicolas M B Brancucci
ABSTRAKT
Plasmodium falciparum accounts for the majority of over 600,000 malaria-associated deaths annually. Parasites resistant to nearly all antimalarials have emerged and the need for drugs with alternative modes of action is thus undoubted. The FK506-binding protein PfFKBP35 has gained attention as a promising drug target due to its high affinity to the macrolide compound FK506 (tacrolimus). Whilst there is considerable interest in targeting PfFKBP35 with small molecules, a genetic validation of this factor as a drug target is missing and its function in parasite biology remains elusive. Here, we show that limiting PfFKBP35 levels are lethal to P. falciparum and result in a delayed death-like phenotype that is characterized by defective ribosome homeostasis and stalled protein synthesis. Our data furthermore suggest that FK506, unlike the action of this drug in model organisms, exerts its antiproliferative activity in a PfFKBP35-independent manner and, using cellular thermal shift assays, we identify putative FK506-targets beyond PfFKBP35. In addition to revealing first insights into the function of PfFKBP35, our results show that FKBP-binding drugs can adopt non-canonical modes of action - with major implications for the development of FK506-derived molecules active against Plasmodium parasites and other eukaryotic pathogens.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Choline chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Anti-Puromycin Antibody, clone 12D10, clone 12D10, from mouse
Informacje o cenach i dostępności nie są obecnie dostępne.
Roche
Anty-GFP, from mouse IgG1κ (clones 7.1 and 13.1)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Informacje o cenach i dostępności nie są obecnie dostępne.