Merck
Chemical composition, antioxidant, and antimicrobial activities of essential oil from pine needle (Cedrus deodara).
Journal of food science (2012-07-05)
Wei-Cai Zeng, Zeng Zhang, Hong Gao, Li-Rong Jia, Qiang He
ABSTRAKT
The chemical composition of essential oil from pine needles (Cedrus deodara) was determined, and its antioxidant and antimicrobial activities were evaluated. Twenty-three components, representing 95.79% of the oil, were identified by gas chromatography mass spectrometry. The main components include α-terpineol (30.2%), linalool (24.47%), limonene (17.01%), anethole (14.57%), caryophyllene (3.14%), and eugenol (2.14%). Pine needle essential oil showed remarkable antioxidant activity in scavenging free radicals, in lipid peroxidation, and in reducing power assays. Moreover, the essential oil revealed strong antimicrobial activity against typical food-borne microorganisms, with minimum inhibitory concentration and minimum bactericidal concentration values of 0.2 to 1.56 and 0.39 to 6.25 μg/mL, respectively. Transmission electron microscope observation ascertained that the bactericidal mechanism of pine needle essential oil may be the induction of cytoplasmic outflow and plasmolysis. These results suggest that the essential oil from pine needles has potential to be used as a natural antioxidant and antimicrobial agent in food processing. The present study provides a theoretical basis for the potential application of essential oil from pine needles (C. deodara) to be used as a natural resource of antioxidant and antimicrobial agents in food industry.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Terpineol, mixture of isomers, 96%, FG
Sigma-Aldrich
(−)−α-Terpineol, natural, ≥96%, FCC, FG
Sigma-Aldrich
(−)-Linalool, ≥95.0% (sum of enantiomers, GC)
Supelco
(−)-Linalool, analytical standard
Sigma-Aldrich
β-Caryophyllene, ≥80%, FCC, FG
Sigma-Aldrich
L-Linalool, natural, ≥95%, FG
Sigma-Aldrich
α-Terpineol, 90%, technical grade
Supelco
(−)-trans-Caryophyllene, analytical standard
Supelco
(+)-α-Terpineol, analytical standard
Sigma-Aldrich
(−)-trans-Caryophyllene, ≥98.0% (sum of enantiomers, GC)
Supelco
trans-Anethole, analytical standard
Sigma-Aldrich
Linalool, 97%
Sigma-Aldrich
trans-Anethole, ≥99%, FCC, FG
Supelco
Linalool, analytical standard
Sigma-Aldrich
Linalool, ≥97%, FCC, FG
Supelco
trans-Anethole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
trans-Anethole, 99%