Merck
  • Simultaneous determination of 3-monochloropropane-1,2-diol and acrylamide in food by gas chromatography-triple quadrupole mass spectrometry with coupled column separation.
Simultaneous determination of 3-monochloropropane-1,2-diol and acrylamide in food by gas chromatography-triple quadrupole mass spectrometry with coupled column separation.
Analytica chimica acta (2012-12-26)
Xiao-min Xu, Hua-li He, Yan Zhu, Liang Feng, Ying Ying, Bai-fen Huang, Hai-tao Shen, Jian-long Han, Yi-ping Ren
ABSTRAKT
Both 3-monochloropropane-1,2-diol (3-MCPD) and acrylamide are contaminants found in heat-processed foods and their related products. A quantitative method was developed for the simultaneous determination of both contaminants in food by gas chromatography-triple quadrupole mass spectrometry (GC-MS/MS). The analytes were purified and extracted by the matrix solid-phase dispersion extraction (MSPDE) technique with Extrelut NT. A coupled column (a 3 m Innowax combined with a 30 m DB-5 ms) was developed to separate both compounds efficiently without derivatization. Triple quadrupole mass spectrometry in multiple reaction monitoring mode (MRM) was applied to suppress matrix interference and obtain good sensitivity in the determination of both analytes. The limit of detection (LOD) in the sample matrix was 5 μg kg(-1) for 3-MCPD or acrylamide. The average recoveries for 3-MCPD and acrylamide in different food matrices were 90.5-107% and 81.9-95.7%, respectively, with the intraday relative standard deviations (RSDs) of 5.6-13.5% and 5.3-13.4%, respectively. The interday RSDs were 6.1-12.6% for 3-MCPD and were 5.0-12.8% for acrylamide. Both contaminants were found in samples of bread, fried chips, fried instant noodles, soy sauce, and instant noodle flavoring. Neither 3-MCPD nor acrylamide was detected in the samples of dairy products (solid or liquid samples) and non-fried instant noodles.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Supelco
Acrylamide solution, 40% in H2O
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder