Merck
Hydrolysis of chickpea proteins with Flavourzyme immobilized on glyoxyl-agarose gels improves functional properties.
Food science and technology international = Ciencia y tecnologia de los alimentos internacional (2013-02-22)
María del Mar Yust, María del Carmen Millán-Linares, Juan María Alcaide-Hidalgo, Francisco Millán, Justo Pedroche
ABSTRAKT
Chickpea protein isolate was hydrolyzed using Flavourzyme immobilized on glyoxyl-agarose beads by multipoint covalent attachment. This Flavourzyme-glyoxyl derivative, produced after 1 h of immobilization at 4 °C followed by 5.5 h at room temperature, presented approximately 51% of the endoprotease activity of Flavourzyme but was around 700 times more stable than soluble enzyme. Chickpea protein hydrolysates ranging from 1% to 10% degree of hydrolysis were produced and their chemical composition was very close to that of protein isolate used as starting material. Solubility, oil absorption, emulsifying activity and stability, and foaming capacity and stability were determined. All protein hydrolysates showed higher solubility than intact proteins, especially at pHs near isoelectric point of native chickpea proteins. Moreover, all hydrolysates had better functional properties, except emulsifying activity, than the original protein isolate.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sepharose 4B, 45-165 μm bead diameter
Sigma-Aldrich
Agarose, Type I, low EEO
Sigma-Aldrich
Agarose, Medium EEO, Molecular Biology
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature, molecular biology grade
Sigma-Aldrich
Agarose, Low EEO
Sigma-Aldrich
Agarose, High EEO
Sigma-Aldrich
Agarose, Low EEO, for Immunoelectrophoresis
Sigma-Aldrich
Agarose, Type II-A, Medium EEO
Sigma-Aldrich
Agarose, Wide range, Molecular Biology
Sigma-Aldrich
Agarose, High EEO
Sigma-Aldrich
Agarose, Special High EEO
Sigma-Aldrich
Agarose, Molecular Biology
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature
Sigma-Aldrich
Agarose, BioReagent, Molecular Biology, low EEO
Sigma-Aldrich
Agarose, High Gelling Temperature
Sigma-Aldrich
Agarose, Type IV, Special High EEO
Sigma-Aldrich
Sepharose 6B, 6% Beaded Agarose, 45-165 μm (wet), fractionation range 10,000-1,000,000 Da (dextrans)
Sigma-Aldrich
Agarose, Type I-A, low EEO
Sigma-Aldrich
Agarose, For pulsed field electrophoresis running gel