Merck
Modulation of cell surface density of carbonic anhydrase IX by shedding of the ectodomain and endocytosis.
Acta virologica (2013-04-23)
M Zatovičová, S Pastoreková
ABSTRAKT
Carbonic anhydrase IX (CA IX) is a cell surface protein frequently present in human tumors but not in the corresponding normal tissues. Expression of CA IX is primarily determined by the strong transcriptional activation of the gene encoding CA IX by hypoxia via the hypoxia-inducible transcription factor HIF-1. However, the ultimate abundance of the CA IX protein on the cell surface can be affected by additional mechanisms, including ectodomain shedding and endocytosis. In this paper, we summarize the basic knowledge on how these processes modulate CA IX expression at the post-translational level. We propose that the regulation of the CA IX protein residence in the plasma membrane can influence its biological function as well as its clinical exploitation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, ≥95% (SDS-PAGE), specific activity ≥3,500 W-A units/mg protein, lyophilized powder
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, BioReagent, suitable for GFC marker
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, For use as a marker in SDS-PAGE
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, non-denaturing PAGE marker
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase I from human erythrocytes
Supelco
Carbonic Anhydrase I from human erythrocytes, Isoelectric focusing marker, pI 6.6
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II human, ≥80%, powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, Isoelectric focusing marker, pI 5.4