Merck
[Alkaline phosphatase activity in blood group B or O secretors is fluctuated by the dinner intake of previous night].
Rinsho byori. The Japanese journal of clinical pathology (2013-07-17)
Makoto Matsushita, Sanae Harajiri, Shiori Tabata, Nobuyasu Yukimasa, Yoshimi Muramoto, Tsugikazu Komoda
ABSTRAKT
We previously reported that two intestinal alkaline phosphatase (IAP) isoforms, high molecular mass IAP (HIAP) and normal molecular mass IAP (NIAP), appear in healthy serum with our Triton-PAGE method for determination of ALP isozymes. In addition, HIAP is chiefly present in blood group B or O secretors, and a large amount of NIAP is secreted into the circulation after high-fat meal in blood group B or O secretors. In the present paper, we investigated the relationship between alkaline phosphatase (ALP) activity in early morning with the patient in a fasted state and the dinner intake of previous night. Two types of dinner were prepared; a low-fat meal (520 kcal), and a high-fat meal (1,040 kcal). Subjects ate the 2 types of dinner on different days. The mean ALP activities at 14 h after high-fat meal ingestion in blood group B or O secretors (n=14) from JSCC and IFCC methods were 8.8% and 5.2% higher than those at 14 h after low-fat meal ingestion in blood group B or O secretors, respectively. The increases in ALP activity between after high-fat meal and low-fat meal were nearly identical to the increases in NIAP activity. These results suggest that a high-fat meal is more likely to affect ALP activity at the early morning with the patient in a fasted state in blood group B or O secretors.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Phosphatase, Alkaline from calf intestinal mucosa, suitable for enzyme immunoassay, solution (clear, colorless), ~2500 U/mg protein (~10 mg/ml)
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host