Merck
Fabrication and characterization of gold nanoflowers formed via chitosan-tripolyphosphate template films for biomedical applications.
Journal of nanoscience and nanotechnology (2013-07-26)
Tran Thi Nhung, Ik-Joong Kang, Sang-Wha Lee
ABSTRAKT
Flowerlike gold nanostructure was facilely prepared by the seed-mediated growth of gold nanoparticles in chitosan-TPP template films. Firstly, chitosan nanoparticles, spontaneously formed by inter and/or intra cross-linking with TPP, were transformed into gel-type of chitosan-TPP films under strong sonication process. After then, gold seeds (1-3 nm) were entrapped in chitosan-TPP films via electrostatic interactions, and the resulting films were subsequently used as a template for growing gold nanoparticles and subsequent formation of anisotropic gold nanoflowers. The size and optical properties of gold nanoflowers were controlled simply by changing the ratio of gold salts to chitosan-TPP template films. Gold nanoflowers consisted of many aggregated nanodots exhibited broad Plasmon-derived absorption bands with strong red-shift into NIR wavelength, which might be a good prospect for SERS and biomedical applications.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chitosan, high molecular weight
Supelco
Sodium ascorbate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium tripolyphosphate, technical grade, 85%
Sigma-Aldrich
Sodium triphosphate pentabasic, purum p.a., ≥98.0% (T)
Sigma-Aldrich
Gold, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Gold, powder, <45 μm, 99.99% trace metals basis
Sigma-Aldrich
Gold, beads, 1-6 mm, 99.999% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, rod, diam. 3.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.997% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.05 mm, 99.99% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)