Merck
  • In vivo and in vitro nuclear magnetic resonance spectroscopy as a tool for investigating hepatobiliary disease: a review of H and P MRS applications.
In vivo and in vitro nuclear magnetic resonance spectroscopy as a tool for investigating hepatobiliary disease: a review of H and P MRS applications.
Liver international : official journal of the International Association for the Study of the Liver (2005-03-23)
Shahid A Khan, I Jane Cox, Gavin Hamilton, Howard C Thomas, Simon D Taylor-Robinson
ABSTRAKT
Nuclear magnetic resonance (NMR) spectroscopy is a non-invasive technique, which allows the study of cellular biochemistry and metabolism. It is a diverse research tool, widely used by biochemists to investigate pathophysiological processes in vitro and, more recently, by physicians to determine disease abnormalities in vivo. This article reviews the basics of the NMR phenomenon and summarises previous research on the hepatobiliary system using both laboratory-based and clinical methodologies. The role of proton and phosphorus-31 ((31)P) NMR spectroscopy in the study of malignant and non-malignant liver disease and studies of bile composition are discussed. In vivo techniques (magnetic resonance spectroscopy, MRS) can be performed as an adjunct to standard MR examination of the liver. Although still primarily a research tool, the in vivo technique provides non-invasive biochemical information on disease severity and holds promise in its use to gauge response to treatment regimens.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-(+)-Lactic acid solution, 30% in H2O (by weight)
SAFC
Glycine
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
L-(+)-Lactic acid solution, meets analytical specification of Ph. Eur., BP, 88-92% total acid basis
Sigma-Aldrich
L-(+)-Lactic acid solution, ~40% in H2O
Sigma-Aldrich
L-(+)-Lactic acid solution, in H2O, ≥85%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine 1 M solution
Supelco
L-(+)-Lactic acid, analytical standard
Sigma-Aldrich
Taurine, ≥98%, FG
SAFC
Taurine
Sigma-Aldrich
L-(+)-Lactic acid, BioXtra, ≥98% (titration)
Sigma-Aldrich
L-(+)-Lactic acid, ≥98%
Supelco
Taurine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Taurine, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
Taurine, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Taurine, ≥99%
Sigma-Aldrich
L-(+)-Lactic acid, 80%