Merck
Polyamine concentrations in pancreatic tissue, serum, and urine of patients with pancreatic cancer.
Pancreas (1990-03-01)
C Löser, U R Fölsch, C Paprotny, W Creutzfeldt
ABSTRAKT
We investigated the total, free, and acetylated polyamine concentrations in pancreatic tissue, serum, and urine of 20 patients with pancreatic cancer, 30 healthy volunteers, and 40 patients with nonmalignant, gastrointestinal diseases by reversed-phase liquid chromatography. Tissue concentrations in carcinoma compared to histologically unaffected pancreas were significantly higher for putrescine, elevated for cadaverine, and nearly identical for spermidine and spermine, while N1-acetylspermidine was detectable in cancer tissue only. With the exception of free spermine in urine and total spermine in serum, all other polyamines were significantly elevated in the urine and serum of cancer patients compared to healthy controls. These data support the concept that polyamines play an important role in rapidly growing tissues. However, nonmalignant gastrointestinal diseases partly showed similar elevations. Because of this low specificity, polyamines are of little value only as diagnostic markers of pancreatic carcinoma. Since polyamine concentrations normalized in patients after curative operation while they were further elevated in patients with tumor relapse or metastases, polyamines might play a clinical role in predicting therapeutic success or indicating relapse of the tumor. A significant linear correlation of polyamine concentrations and the size of the tumor was found while a significant correlation to CEA, CA 19-9, and CA 125 or the presence of organ metastases did not exist.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg
Sigma-Aldrich
Retinol, ≥95.0% (HPLC), ~2700 U/mg
Sigma-Aldrich
1,4-Diaminobutane, 99%
Supelco
4-Androstene-3,17-dione, VETRANAL®, analytical standard
Sigma-Aldrich
Cadaverine, purum, ≥97.0% (GC)
Sigma-Aldrich
Cadaverine, 95%
Supelco
Androstenedione solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Supelco
Linoleic acid, analytical standard
Sigma-Aldrich
cis-4,7,10,13,16,19-Docosahexaenoic acid, ≥98%
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g