Merck
  • Investigations into acrylamide precursors in sterilized table olives: evidence of a peptic fraction being responsible for acrylamide formation.
Investigations into acrylamide precursors in sterilized table olives: evidence of a peptic fraction being responsible for acrylamide formation.
Food chemistry (2013-06-26)
Francisco Javier Casado, Alfredo Montaño, Dietrich Spitzner, Reinhold Carle
ABSTRAKT
Formation of acrylamide from commercial model peptides containing protein-bound aspartic acid, alanine and methionine, respectively, at 200°C and different times in the absence of any carbonyl sources, was demonstrated by HPLC-MS/MS analyses. Further experiments using a more complex model system based on olive water, i.e., the aqueous fraction of olive pulp from untreated and lye-treated green olives, were performed. After partial fractionation of olive water by solid-phase extraction, only peptides/proteins containing fractions, being devoid of free asparagine, generated significant amounts of acrylamide during less harsh heat treatment (121°C for 30min). In contrast, acrylamide was not detected after heating the same fraction under identical thermal conditions when previously subjected to acid hydrolysis. Consistently, significant amounts of acrylamide were released after heating the albuminous precipitate resulting from acetone precipitation of olive water. These results strongly support the role of peptides/proteins as precursors of acrylamide formation in sterilized olives.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Supelco
Acrylamide solution, 40% in H2O