Merck
Do rhizospheric processes linked to P nutrition participate in U absorption by Lupinus albus grown in hydroponics?
Journal of environmental radioactivity (2013-07-09)
Antoine Tailliez, Sylvie Pierrisnard, Virginie Camilleri, Catherine Keller, Pascale Henner
ABSTRAKT
Phosphate (P) is an essential element for plant development but is generally present in limiting amount in the soil solution. Plant species have developed different mechanisms promoting the solubilization of this element in soils to ensure a sufficient supply for their growth. One of these mechanisms is based on the ability of certain species such as L. albus to exude large amounts of citrate through specific tertiary roots called cluster-roots. Uranium (U) is an ubiquitous contaminant known firstly for its chemical toxicity and secondly for its high affinity for P with which it forms low-soluble complexes in soils. We highlight the effects of P-U interaction on the physiology of L. albus and particularly on citrate exudation, and the impact of this root process on the phytoavailability of U and its accumulation in plants in a hydroponic study. Different levels of P (1 and 100 μM) and U (0 and 20 μM) have been tested. Our results show no toxicity of U on the development of L. albus with an adequate P supply, whereas the effects of P starvation are amplified by the presence of U in the growth medium, except for the production of cluster-roots. Citrate exudation is totally inhibited by U in a low-P environment whereas it increases in the presence of U when its toxicity is lowered by the addition of P. The differences observed in terms of toxicity and accumulation are partly explained by the microphotographs obtained by electron microscopy (TEM-EDX): in the absence of P, U penetrates deep into the roots and causes lethal damages, whereas in presence of P, we observe the formation of U-P complexes which limit the internalization of the pollutant and so its toxicity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, 99%
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland