Merck
Förster resonance energy transfer beyond 10 nm: exploiting the triplet state kinetics of organic fluorophores.
The journal of physical chemistry. B (2011-09-21)
Heike Hevekerl, Thiemo Spielmann, Andriy Chmyrov, Jerker Widengren
ABSTRAKT
Inter- or intramolecular distances of biomolecules can be studied by Förster resonance energy transfer (FRET). For most FRET methods, the observable range of distances is limited to 1-10 nm, and the labeling efficiency has to be controlled carefully to obtain accurate distance determinations, especially for intensity-based methods. In this study, we exploit the triplet state of the acceptor fluorophore as a FRET readout using fluorescence correlation spectroscopy and transient state monitoring. The influence of donor fluorescence leaking into the acceptor channel is minimized by a novel suppression algorithm for spectral bleed-through, thereby tolerating a high excess (up to 100-fold) of donor-only labeled samples. The suppression algorithm and the high sensitivity of the triplet state to small changes in the fluorophore excitation rate make it possible to extend the observable range of FRET efficiencies by up to 50% in the presence of large donor-only populations. Given this increased range of FRET efficiencies, its compatibility with organic fluorophores, and the low requirements on the labeling efficiency and instrumentation, we foresee that this approach will be attractive for in vitro and in vivo FRET-based spectroscopy and imaging.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Atto 488 amine, BioReagent, ≥90% (HPLC), suitable for fluorescence
Sigma-Aldrich
Atto 488 azide, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 488 maleimide, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 488-Biotin, BioReagent, suitable for fluorescence, ≥90.0% (HPLC)
Sigma-Aldrich
Atto 488 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 488, BioReagent, suitable for fluorescence, ≥90% (HPLC)