Merck
A pilot study of pepsin in tracheal and oral secretions.
American journal of critical care : an official publication, American Association of Critical-Care Nurses (2013-09-03)
Marilyn Schallom, Sally M Tricomi, Yie-Hwa Chang, Norma A Metheny
ABSTRAKT
Because reflux of gastric juice into the oropharynx must precede its aspiration into the lungs, it is reasonable to hypothesize that the detection of pepsin (the major gastric enzyme in gastric juice) in oral secretions may provide a relatively noninvasive method of predicting risk for aspiration. To describe the incidence of pepsin in oral and tracheal secretions collected concurrently from a sample of 50 gastric-fed patients undergoing mechanical ventilation. An exploratory descriptive design with a convenience sample from 4 medical and surgical intensive care units. An oral secretion and a tracheal secretion were collected concurrently from each patient (yielding a sample of 50 oral and 50 tracheal secretions). The tracheal secretions were obtained via the inline suction system with an attached sputum trap; oral secretions were obtained via a Yankauer suction tip with an attached sputum trap. All specimens were assayed for pepsin by the Western blot method. Oral secretions from 10 patients (20%) and tracheal secretions from 2 patients (4%) were pepsin-positive. Both patients with pepsin-positive tracheal secretions also had pepsin-positive oral secretions. Pepsin was not found in the tracheal secretions from the remaining 8 patients with pepsin-positive oral secretions. Although reflux of gastric juice into the oropharynx must precede its aspiration into the lungs, individual reflux events do not necessarily lead to aspiration. Thus, it is reasonable that we found pepsin 5 times more often in oral secretions than in tracheal secretions.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
Pepsin−Agarose from porcine gastric mucosa, lyophilized powder, ≥30 units/mg dry solid
Sigma-Aldrich
Pepsin from porcine gastric mucosa, tested according to Ph. Eur.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, slightly beige, 1200-2400 U/mg
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, slightly beige, ≥500 U/mg
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥400 units/mg protein
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥2,500 units/mg protein (E1%/280)
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid
Sigma-Aldrich
Pepsin from porcine gastric mucosa, Suitable for manufacturing of diagnostic kits and reagents, lyophilized powder, ≥3200 units/mg protein