Merck
Expression levels of serine/glycine metabolism-related proteins in triple negative breast cancer tissues.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2014-01-07)
Songmi Noh, Do Hee Kim, Woo Hee Jung, Ja Seung Koo
ABSTRAKT
To evaluate the expression levels of serine/glycine metabolism-related proteins (PHGDH, PSAT, PSPH, SHMT, and GLDC) in six different subtypes of triple negative breast cancer (TNBC) patients and gain insight into their implications. Formalin-fixed, paraffin-embedded tissues from 129 TNBC patients were assembled into tissue microarrays. Immunohistochemical staining was performed for serine/glycine metabolism-related proteins (PHGDH, PSAT, PSPH, SHMT, and GLDC) and surrogate immunohistochemical markers (CK5/6, EGFR, claudin 3, claudin 4, claudin 7, E-cadherin, STAT1, interleukin-8 [IL-8], AR, and GGT-1) for identifying the molecular subtype of TNBC. TNBC subtype classifications included the following: basal-like (CK5/6-positive and/or EGFR-positive), molecular apocrine (AR-positive and/or GGT-1-positive), claudin-low (claudin 3-, claudin 4-, claudin 7-negative and/or E-cadherin-negative), immune-related (IL-8-negative and stromal STAT1-positive), mixed (features from two or more of the four subtypes), and null (no features from any of the four subtypes). Tissues from basal marker-positive patients showed increased expression levels of tumoral PHGDH compared with those from basal marker-negative patients (p = 0.029); lack of stromal SHMT1 expression was significantly correlated with T stage (p = 0.016). Multivariate Cox analysis revealed that a lack of stromal SHMT1 expression was an independent prognostic factor for predicting a shorter disease-free survival period (hazard ratio 4.002, 95 % confidence interval [CI] 1.077-14.83, p = 0.038); furthermore, a lack of tumoral PHGDH expression was predictive of a shorter overall survival rate (hazard ratio 3.053, 95 % CI 1.002-9.305, p = 0.050). In conclusion, the most abundantly expressed serine/glycine metabolism-related protein in basal-like TNBC tissues was tumoral PHGDH, and expression levels of stromal SHMT1 and tumoral PHGDH were inversely correlated with clinical prognostic factors. Also, this study is the first to assess serine/glycine relationships at the protein level in regards to clinical outcomes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Glycine, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Supelco
Glycine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
SAFC
Glycine
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Serine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glycine 1 M solution
Supelco
L-Serine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Serine, ReagentPlus®, ≥99% (HPLC)
Supelco
L-Serine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Serine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Serine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Glycine hydrochloride, ≥99% (HPLC)