Merck
  • Osteoblastic differentiation and mineralization ability of periosteum-derived cells compared with bone marrow and calvaria-derived cells.
Osteoblastic differentiation and mineralization ability of periosteum-derived cells compared with bone marrow and calvaria-derived cells.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons (2014-02-01)
Juan Marcelo Rosales-Rocabado, Masaru Kaku, Megumi Kitami, Yosuke Akiba, Katsumi Uoshima
ABSTRAKT
Clinically, bone marrow stromal cells (BMCs) are the most common source of osteoprogenitor cells. Its harvest process, however, is invasive to patients. Previous reports have shown the potential advantages of using periosteum-derived cells (PDCs) as a source of cell-based transplant therapy. The objective of our study was to characterize the osteoblastic differentiation and mineralization ability of PDCs versus BMCs and osteoblasts (OBs). BMCs, OBs, and PDCs were isolated from 4-week-old male Wistar rats. To characterize the differentiation ability of the cells, MTS assay, alkaline phosphatase (ALP) activity staining, picrosirius red staining, and alizarin red staining were performed. Immunohistochemistry was performed on paraffin sections of calvarial periosteum to determine the presence of mesenchymal stem cells. PDCs showed the greatest proliferation rate compared with BMCs and OBs. Matured collagenous matrix formation was observed in PDCs and BMCs. ALP-positive cells and in vitro mineralization were evident in all cell types analyzed; however, that of PDCs was not comparable to that of the OBs and BMCs. Immunohistochemistry revealed the presence of STRO-1-and CD105-positive cells in the cambium layer of the periosteum. PDCs have remarkable proliferative ability, but contain only a small population of osteogenic cells compared with BMCs and OBs. Although cell activity can be affected by various factors, such as age, culture condition, additives, and so forth, PDCs are likely not the source of OBs, although they might provide matrices that indirectly aid in bone formation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Alizarin, Dye content 97 %
Sigma-Aldrich
Phosphatase, Alkaline from calf intestinal mucosa, suitable for enzyme immunoassay, solution (clear, colorless), ~2500 U/mg protein (~10 mg/ml)
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host