Merck
  • Maternal nicotinamide supplementation causes global DNA hypomethylation, uracil hypo-incorporation and gene expression changes in fetal rats.
Maternal nicotinamide supplementation causes global DNA hypomethylation, uracil hypo-incorporation and gene expression changes in fetal rats.
The British journal of nutrition (2014-02-11)
Yan-Jie Tian, Ning Luo, Na-Na Chen, Yong-Zhi Lun, Xin-Yi Gu, Zhi Li, Qiang Ma, Shi-Sheng Zhou
ABSTRAKT
Recent evidence shows that excess nicotinamide can cause epigenetic changes in developing rats. The aim of the present study was to investigate the effects of maternal nicotinamide supplementation on the fetus. Female rats were randomised into four groups fed a standard chow diet (control group) or diets supplemented with 1 g/kg of nicotinamide (low-dose group), 4 g/kg of nicotinamide (high-dose group) or 4 g/kg of nicotinamide plus 2 g/kg of betaine (betaine group) for 14-16 d before mating and throughout the study. Fetal tissue samples were collected on the 20th day of pregnancy. Compared with the control group, the high-dose group had a higher fetal death rate, and the average fetal body weight was higher in the low-dose group but lower in the high-dose group. Nicotinamide supplementation led to a decrease in placental and fetal hepatic genomic DNA methylation and genomic uracil contents (a factor modifying DNA for diversity) in the placenta and fetal liver and brain, which could be completely or partially prevented by betaine. Moreover, nicotinamide supplementation induced tissue-specific alterations in the mRNA expression of the genes encoding nicotinamide N-methyltransferase, DNA methyltransferase 1, catalase and tumour protein p53 in the placenta and fetal liver. High-dose nicotinamide supplementation increased fetal hepatic α-fetoprotein mRNA level, which was prevented by betaine supplementation. It is concluded that maternal nicotinamide supplementation can induce changes in fetal epigenetic modification and DNA base composition. The present study raises the concern that maternal nicotinamide supplementation may play a role in the development of epigenetic-related diseases in the offspring.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
Sigma-Aldrich
Uracil, ≥99.0%
Sigma-Aldrich
Uracil, BioReagent, suitable for cell culture
Sigma-Aldrich
Betaine hydrochloride, ≥99%
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Niacinamide, meets USP testing specifications
USP
Niacynamid, United States Pharmacopeia (USP) Reference Standard
Supelco
Nicotinamide (Vitamin B3) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Fluorouracil impurity C, European Pharmacopoeia (EP) Reference Standard